Learn More
Octanal, 98%
Description
Octanal is an aromatic aldehyde often found in citrus oils. It is used commercially as a component in perfumes and in flavor production for the food industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Octanal is an aromatic aldehyde often found in citrus oils. It is used commercially as a component in perfumes and in flavor production for the food industry.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Handle and store under inert gas. Air and moisture sensitive. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 12°C to 15°C |
| Density | 0.821 |
| Boiling Point | 169°C to 171°C |
| Flash Point | 56°C (133°F) |
| Refractive Index | 1.4195 |
| Linear Formula | CH3(CH2)6CHO |
| Quantity | 50 g |
| UN Number | UN1191 |
| Sensitivity | Air Sensitive |
| Merck Index | 14,1766 |
| Show More |
Safety and Handling
| Hazard Category | H226-H315-H319 |
| Hazard Statement | GHS H Statement H226-H315-H319-H335 Flammable liquid and vapor. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P374-P380-P390-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: OCTYL ALDEHYDES |
| EINECSNumber | 204-683-8 |
| RTECSNumber | RG7780000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.