Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octanoic acid, 98+%

Catalog No. aaa111490d
Click to view available options
Quantity:
10,000 mL
25 mL
2500 mL
500 mL

CAS: 124-07-2 | C8H16O2 | 144.214 g/mol

Octanoic acid is used in the production of esters, which finds application in perfumery and also dyes manufacture. It is used as a food contact surface sanitizer in commercial food handling, as a dietary supplement and as disinfectant in health care facilities, recreational facilities, livestock premises and industrial facilities. Its acid chloride is used in the synthesis of perfluorooctanoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Octanoic acid is used in the production of esters, which finds application in perfumery and also dyes manufacture. It is used as a food contact surface sanitizer in commercial food handling, as a dietary supplement and as disinfectant in health care facilities, recreational facilities, livestock premises and industrial facilities. Its acid chloride is used in the synthesis of perfluorooctanoic acid.

Solubility
Misicible with alcohol, chloroform, ether, carbon disulfide, petroleum ether and acetonitrile.Slightly miscible with water.

Notes
Incompatible with bases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-07-2
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.214
MDL Number MFCD00004429
InChI Key WWZKQHOCKIZLMA-UHFFFAOYSA-N
Synonym caprylic acid, n-octanoic acid, octylic acid, n-caprylic acid, octoic acid, n-octylic acid, n-octoic acid, neo-fat 8, 1-heptanecarboxylic acid, enantic acid
PubChem CID 379
ChEBI CHEBI:28837
IUPAC Name octanoic acid
SMILES CCCCCCCC(=O)O

Specifications

Melting Point 16°C to 17°C
Density 0.91
pH 4
Boiling Point 236°C to 238°C
Flash Point 132°C (269°F)
Odor Rancid
Assay Percent Range >98%
Refractive Index 1.4275
Linear Formula CH3(CH2)6CO2H
Quantity 10,000 mL
UN Number UN3265
Beilstein 1747180
Merck Index 14,1765
Solubility Information Misicible with alcohol,chloroform,ether,carbon disulfide,petroleum ether and acetonitrile.Slightly miscible with water.
Formula Weight 144.21
Percent Purity ≥98%
Chemical Name or Material Octanoic acid
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 204-677-5
RTECSNumber RH0175000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.