Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Glycine tert butyl ester hydrochloride, 99%

Catalog No. AC297020025
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle

CAS: 27532-96-3 | C6H14NO2 | 132.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27532-96-3
Molecular Formula C6H14NO2
Molecular Weight (g/mol) 132.18
MDL Number MFCD00058255
InChI Key SJMDMGHPMLKLHQ-UHFFFAOYSA-O
Synonym h-gly-otbu.hcl, tert-butyl 2-aminoacetate hydrochloride, glycine tert-butyl ester hydrochloride, glycine tert butyl ester hydrochloride, glycine tert-butyl ester hcl, glycine t-butyl ester hydrochloride, tert-butyl glycine hydrochloride, t-butyl glycinate, tert-butyl aminoacetate hydrochloride, glycine tert-butylester hydrochloride
PubChem CID 2725036
IUPAC Name tert-butyl 2-aminoacetate;hydrochloride
SMILES CC(C)(C)OC(=O)C[NH3+]

Specifications

Melting Point 138°C to 142°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula H2NCH2C(O)OC(CH3)3·HCl
Quantity 2.5 g
Formula Weight 167.63
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Glycine tert butyl ester hydrochloride, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.