Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octanoyl Chloride 99%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass bottle
111-64-8
C8H15ClO
162.66
MFCD00000766
REEZZSHJLXOIHL-UHFFFAOYSA-N
capryloyl chloride,caprylyl chloride,n-octanoyl chloride,caprylic acid chloride,octanoic acid chloride,octanoic chloride,octanoylchloride,ccris 5990,caprylic chloride,octanoyl-chloride
8124
octanoyl chloride
CCCCCCCC(=O)Cl
111-64-8
C8H15ClO
162.66
MFCD00000766
REEZZSHJLXOIHL-UHFFFAOYSA-N
capryloyl chloride,caprylyl chloride,n-octanoyl chloride,caprylic acid chloride,octanoic acid chloride,octanoic chloride,octanoylchloride,ccris 5990,caprylic chloride,octanoyl-chloride
8124
octanoyl chloride
CCCCCCCC(=O)Cl
Viscosity 1.51 mPa.s (20°C)
Specific Gravity 0.95
Refractive Index 1.4340 to 1.4360
Melting Point -63.0°C
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 2.5kg
Flash Point 75°C
Color Undesignated
Acidity 1% max.
Density 0.9500g/mL
Boiling Point 195.0°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula CH3(CH2)6COCl
Beilstein 02, 348
Solubility Information Solubility in water: reacts. Other solubilities: reacts with alcohol, miscible with most common organic solvents, soluble in ether
Formula Weight 162.66
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Octanoyl chloride, 99%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 203-891-6

RTECSNumber : RH1570000

TSCA : TSCA