Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Octanoyl-L-carnitine, ≥97.0% (HPLC), MilliporeSigma™ Supelco™
Click to view available options
Quantity:
10 mg
Specifications
Specifications
CAS | 25243-95-2 |
Linear Formula | C15H29 NO4 |
Synonym | (2 R)-3-Carboxy-N,N,N-trimethyl-2-[(1-oxooctyl)oxy]-1-propanaminium inner salt; L-Carnitine octanoyl ester; C8-Carnitine |
Type | Analytical Standard |
Quantity | 10 mg |
Formula Weight | 287.40 |
Percent Purity | ≥97.0% (HPLC) |
Grade | Analytical Standard |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction