Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octinoxate, USP, 95-105%, Spectrum™ Chemical
SDP

Catalog No. 18604868 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
500 g
2.5 kg
20 kg
Packaging:
Amber Glass Bottle
Steel Pail

O1123, 5466-77-3, C18H26O3

Spectrum™ Chemical Octinoxate, USP absorbs ultraviolet radiation in the UVB range and is used in sunscreens and other cosmetics to protect from skin damage. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 5466-77-3
Molecular Formula C18H26O3
Molecular Weight (g/mol) 290.40
MDL Number MFCD00072582
InChI Key YBGZDTIWKVFICR-UHFFFAOYNA-N
IUPAC Name 2-ethylhexyl 3-(4-methoxyphenyl)prop-2-enoate
SMILES CCCCC(CC)COC(=O)C=CC1=CC=C(OC)C=C1

Specifications

Acidity To pass test
CAS Max % 1
Packaging Steel Pail
Refractive Index 1.542-1.548
Quantity 20 kg
Specific Gravity 1.005-1.013
Percent Purity 95 to 105%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.