Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Octisalate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
1 g
10 g
Description
Octisalate is a sunscreen agent that acts as a UV filter for UVB (280-315 nm) radiation.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Pharmaceutical (small molecule)

Specifications
Specifications
CAS | 118-60-5 |
Density | 1.014 g/mL (at 25°C) |
Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available. |
Physical Form | Neat |
Linear Formula | (HO)C6H4 CO2 CH2 CH(C2H5)(CH2)3 CH3 |
MDL Number | MFCD00053300 |
Boiling Point | 189°C to 190°C (21 mmHg) |
Synonym | 2-Ethylhexyl salicylate; Octisalate; Octyl salicylate |
Type | Certified Reference Material |
Quantity | 1 g |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction