Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OCTN1/SLC22A4 Antibody (892147), PE/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB10985PECY7
Description
OCTN1/SLC22A4 Monoclonal antibody specifically detects OCTN1/SLC22A4 in Human samples. It is validated for Flow CytometrySpecifications
OCTN1/SLC22A4 | |
Monoclonal | |
PE-Cyanine7 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
6583 | |
Store at 4C in the dark. Do not freeze. | |
IgG2a |
Flow Cytometry | |
892147 | |
Flow Cytometry | |
Ergothioneine transporter, ET transporter, ETT, MGC34546, MGC40524, OCTN1integral membrane transport protein, Organic cation/carnitine transporter 1, solute carrier family 22 (organic cation/ergothioneine transporter), member 4, solute carrier family 22 member 4, UT2H | |
NS0 mouse myeloma cell line transfected with human SLC22A4, Accession # Q9H015 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction