Learn More
OCTN2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Supplier:  Proteintech Group Inc 163311AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-0876
Only null left
Add to Cart
Description
Description
Carnitine (b-hydroxy-g-trimethylaminobutyrate) is a small, highly polar compound that aids in the b-oxidation of long-chain fatty acids. Organic cation/carnitine transporters (OCTN) assist in the elimination of cationic compounds, including xenobiotics, and transport carnitine for reabsorption in the kidney. Similar to organic cation transporters (OCT), OCTN proteins localize to the plasma membrane of epithelial cells. OCTN1 is expressed in kidney, trachea, bone marrow and fetal liver. OCTN2 is abundantly expressed in kidney, skeletal muscle, placenta and heart. OCTN3 is strongly expressed in testis and weakly expressed in kidney. The gene encoding human OCTN1 maps to chromosome 5 and the gene encoding human OCTN2 maps to chromosome 5q31. Mutations in the gene encoding OCTN2 leads to systemic carnitine deficiency (SCD), an autosomal recessive disorder characterized by cardiomyopathy, skeletal myopathy, lethargy, hypoglycemia and hyperammonemia.
Specifications
Specifications
OCTN2
Polyclonal
Unconjugated
SLC22A5
CDSP, OCTN2, OCTN2VT, SLC22A5
Rabbit
Antigen Affinity Chromatography
RUO
20520, 29726, 6584
-20°C
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O70594, O76082, Q9Z0E8
SLC22A5
OCTN2 Fusion Protein Ag9450
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.