Learn More
Epreida™ Octospot™ Cell Preparation System Accessories
Shop All Epredia Products
Click to view available options
For Use With (Equipment):
Epredia Cytospin
Description
- Cytocentrifuge cells from 8-well microliter strips directly onto a microscope slide (96 spots per spin); turn eight slides into one
- Incorporates an 8-well strip, an 8-well slide, a holding block and a filter card assembled in standard stainless steel
- Cytoclip Solvent-resistant slides retain separate reagents in individual wells to prevent cross-contamination
Specifications
Specifications
| Type | Thick Filter Cards |
| For Use With (Equipment) | Epredia Cytospin |
| Disposable | Single-use |