Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octyl Aldehyde 99%, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
Packaging:
Glass bottle
124-13-0
C8H16O
128.21
MFCD00007029
NUJGJRNETVAIRJ-UHFFFAOYSA-N
caprylic aldehyde,caprylaldehyde,n-octanal,1-octanal,n-octyl aldehyde,n-octaldehyde,n-caprylaldehyde,octanaldehyde,aldehyde c-8,n-octylal
454
CHEBI:17935
octanal
CCCCCCCC=O
124-13-0
C8H16O
128.21
MFCD00007029
NUJGJRNETVAIRJ-UHFFFAOYSA-N
caprylic aldehyde,caprylaldehyde,n-octanal,1-octanal,n-octyl aldehyde,n-octaldehyde,n-caprylaldehyde,octanaldehyde,aldehyde c-8,n-octylal
454
CHEBI:17935
octanal
CCCCCCCC=O
Density 0.8210g/mL
Specific Gravity 0.821
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4170 to 1.4200
Melting Point 12.0°C to 15.0°C
Color Colorless to Yellow
Quantity 100mL
Boiling Point 171.0°C
Flash Point 51°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula CH3(CH2)6CHO
Beilstein 01, 704
Merck Index 15, 1768
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol and mineral oil, insoluble in glycerol
Formula Weight 128.21
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Octyl aldehyde, 99%
Show More Show Less

GHS H Statement:
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 204-683-8

RTECSNumber : RG7780000

TSCA : TSCA