Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octyl sulfate sodium salt, 99.8%, MP Biomedicals™

Catalog No. ICN15598001 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15598001 1 g
ICN15598005 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15598001 Supplier MP Biomedicals, Inc Supplier No. 0215598001

Chemical Identifiers

CAS 142-31-4
Molecular Formula C8H17NaO4S
Molecular Weight (g/mol) 232.27
InChI Key WFRKJMRGXGWHBM-UHFFFAOYSA-M
Synonym sodium octyl sulfate, sodium n-octyl sulfate, sodium octyl sulphate, sipex ols, cycloryl os, duponol 80, sodium capryl sulfate, octyl sodium sulfate, sulfuric acid, monooctyl ester, sodium salt, octyl sulfate sodium salt
PubChem CID 2735107
IUPAC Name sodium;octyl sulfate
SMILES CCCCCCCCOS(=O)(=O)[O-].[Na+]

Specifications

Color White
Physical Form Crystalline Powder
Quantity 1 g
Assay Percent Range 99.8%
Formula Weight 232.3
Chemical Name or Material Octyl sulfate sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.