Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octylmagnesium bromide, 2M solution in diethyl ether,AcroSeal™

Catalog No. p-4089840
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 17049-49-9 | C8H17BrMg | 217.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17049-49-9, 60-29-7
Molecular Formula C8H17BrMg
Molecular Weight (g/mol) 217.43
MDL Number MFCD00191566
InChI Key ZDUZWGUHIRQPGP-UHFFFAOYSA-M
Synonym bromooctyl magnesium, n-octylmagnesium bromide, magnesium, bromooctyl, octylmagnesium bromide, n-octyl magnesium bromide, n-octylmagnesiumbromide, octyl magnesium bromide, c8h17mgbr, 1-octylmagnesium bromide, octylmagnesium bromide solution, grignard reagent
PubChem CID 86923
IUPAC Name bromo(octyl)magnesium
SMILES CCCCCCCC[Mg]Br

Specifications

Boiling Point 35.0°C
Color Yellow to Brown
CAS Min % 42.99, 50.22
CAS Max % 49.78, 57.01
Physical Form Liquid
Quantity 100 mL
Linear Formula CH3(CH2)7MgBr
Flash Point −35°C
Formula Weight 217.43
Concentration or Composition (by Analyte or Components) 1.90 to 2.20M
Packaging AcroSeal™ Glass bottle
Specific Gravity 0.961
Density 0.9610g/mL
Chemical Name or Material Octylmagnesium bromide, 2M solution in diethyl ether
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause drowsiness or dizziness.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Extremely flammable liquid and vapor.
May form explosive peroxides.
Reacts violently with water.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.