Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ OD1

Click to view available options
Quantity:
10 μg
Description
OD1 is a potent rat Nav1.7, human Nav1.4 and rat Nav1.6 channel activator (EC50 values are 7, 10 and 47 nM, respectively). Exhibits minimal activation at mammalian Nav1.2, Nav1.3 and Nav1.5 (EC50 values >3 μM). Inhibits fast inactivation on all channels. Increases peak currents at all voltages and stimulates a persistent Na+ current at hNav1.7 channel. Increases hyperpolarization at Nav1.4 and Nav1.6 channels. Induces spontaneous pain in vivo.
Specifications
Specifications
Purity | 95% |
Content And Storage | Store at -20°C |
Description | Voltage-gated Sodium Channel Blockers |
Molecular Weight (g/mol) | Observed M.W.: 7206.1 |
Quantity | 10 μg |
Structural Form | C308H466N90O95S8 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction