Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ODAM Monoclonal Antibody (2F8)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA524467
Description
Peptide Sequence: MPYVFSFKMP QEQGQMFQYY PVYMVLPWEQ PQQTVPRSPQ QTRQQQYEEQ IPFYAQFGYI PQLAEPAISG GQQQLAFDPQ LGTAPEIAVM STGEEIPYLQ KEAINFRHDS AGVFMPSTSP KPSTTNVFTS AVDQTITPEL PEEKDKTDSL REP.
Tooth-associated epithelia protein that probably plays a role in odontogenesis, the complex process that results in the initiation and generation of the tooth. May be incorporated in the enamel matrix at the end of mineralization process. Involved in the induction of RHOA activity via interaction with ARHGEF and expression of downstream factors such as ROCK. Plays a role in attachment of the junctional epithelium to the tooth surface.
Specifications
ODAM | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
A1E959 | |
ODAM | |
ODAM (AAH17796.1, 1 a.a. ~ 153 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Western Blot | |
2F8 | |
Unconjugated | |
ODAM | |
APIN; ODAM; Odontogenic ameloblast-associated protein; odontogenic, ameloblast asssociated | |
Mouse | |
Protein A | |
RUO | |
54959 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction