Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ODAM Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5139793
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5139793 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5139793 Supplier Invitrogen™ Supplier No. PA5139793
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: IPQRLMSASN SNELLLNLNN GQLLPLQLQG PLNSWIPPFS GILQQQQQAQ IPGLSQFSLS ALDQFAGLLP NQIPLTGEAS FAQGA Highest antigen sequence indentity to the following orthologs: Mouse - 71%, Rat - 70%.

Tooth-associated epithelia protein that probably plays a role in odontogenesis, the complex process that results in the initiation and generation of the tooth. May be incorporated in the enamel matrix at the end of mineralization process. Involved in the induction of RHOA activity via interaction with ARHGEF and expression of downstream factors such as ROCK. Plays a role in attachment of the junctional epithelium to the tooth surface.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ODAM
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ODAM
Gene Accession No. A1E959
Gene Alias APIN; ODAM; Odontogenic ameloblast-associated protein; odontogenic, ameloblast asssociated
Gene Symbols ODAM
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ODAM. Recombinant protein control fragment (Product #RP-108818).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54959
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.