Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ODC1 Monoclonal Antibody (3F293)
GREENER_CHOICE

Catalog No. PIMA125811
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA125811 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA125811 Supplier Invitrogen™ Supplier No. MA125811
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Recommended positive controls: Epithelial cells in normal placenta or prostate carcinoma. MA1-25811 is expected to cross react with all known eukaryotic ODC proteins due to sequence homology.

Ornithine decarboxylase catalyzes the conversion of ornithine to putrescine in the first and apparently rate-limiting step in polyamine biosynthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ODC1
Applications Immunohistochemistry
Classification Monoclonal
Clone 3F293
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Odc1
Gene Accession No. P00860, P09057, P11926
Gene Alias MGC103389; Odc; ODC1; Ornithine decarboxylase; ornithine decarboxylase 1; ornithine decarboxylase, structural 1; Ornitine decarboxylase
Gene Symbols Odc1
Host Species Mouse
Immunogen Synthetic hexadecapeptide (aa345-360, KPDEKYYSSSIWGPTC) from rat ODC. Epitope: aa355-360 (-IWGPTC-).
Purification Method Protein G
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18263, 24609, 4953
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.