Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ODC1 Monoclonal Antibody (MP16-2), Invitrogen™

Catalog No. PIMA513485
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA513485 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA513485 Supplier Thermo Scientific Supplier No. MA513485
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-13485 targets Ornithine Decarboxylase in IHC (F) applications and shows reactivity with Human, mouse, and Rat samples. The MA5-13485 immunogen is a synthetic hexadecapeptide (KPDEKYYSSSIWGPTC) (aa345-360) from rat ODC.

Ornithine decarboxylase catalyzes the conversion of ornithine to putrescine in the first and apparently rate-limiting step in polyamine biosynthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ODC1
Applications Immunohistochemistry (Frozen)
Classification Monoclonal
Clone MP16-2
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene Odc1
Gene Accession No. P00860, P09057, P11926
Gene Alias MGC103389; Odc; ODC1; Ornithine decarboxylase; ornithine decarboxylase 1; ornithine decarboxylase, structural 1; Ornitine decarboxylase
Gene Symbols Odc1
Host Species Mouse
Immunogen A synthetic hexadecapeptide (KPDEKYYSSSIWGPTC) (aa345-360) from rat ODC
Purification Method Protein G
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18263, 24609, 4953
Target Species Human, Mouse, Rat
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.