Learn More
ODC1 Mouse anti-Human, Clone: OTI1G6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF501286

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Ornithine decarboxylase catalyzes the conversion of ornithine to putrescine in the first and apparently rate-limiting step in polyamine biosynthesis.Specifications
ODC1 | |
Monoclonal | |
Unconjugated | |
ODC1 | |
ODC | |
Mouse | |
Affinity Chromatography | |
RUO | |
4953 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI1G6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P11926 | |
ODC1 | |
Full length human recombit protein of human ODC1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.