Learn More
Invitrogen™ ODC1 Polyclonal Antibody

Description
Immunogen sequence: MNNFGNEEFD CHFLDEGFTA KDILDQKINE VSSSDDKDAF YVADLGDILK KHLRWLKALP RVTPFYAVKC NDSKAIVKTL AATGTGFDCA SKTEIQLVQS LGVPPERIIY ANPCKQVSQI KYAANNGVQM MTFDSEVELM KVARAHPKAK LVLRIATDDS KAVCRLSVKF GATLRTSRLL LERAKELNID VVGVSFHVGS GCTDPETFVQ AISDARCVFD MGAEVGFSMY LLDIGGGFPG SEDVKLKFEE ITGVINPALD KYFPSDSGVR IIAEPGRYYV; Positive Samples: HeLa, THP-1, Mouse kidney, Mouse skeletal muscle.
Specifications
Specifications
| Antigen | ODC1 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.77 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 |
| Gene | Odc1 |
| Gene Accession No. | P00860, P09057, P11926 |
| Gene Alias | MGC103389; Odc; ODC1; Ornithine decarboxylase; ornithine decarboxylase 1; ornithine decarboxylase, structural 1; Ornitine decarboxylase |
| Gene Symbols | Odc1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.