Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ODC1 Rabbit anti-Human, Mouse, Rat, Unconjugated, Clone: ODC1/2878R, Abnova™

Catalog No. 89339576 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-576 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-576 Supplier Abnova Corporation Supplier No. RAB00597
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

ODC1 Monoclonal antibody specifically detects ODC1 in Human, Mouse, Rat samples. It is validated for IHC-P.

Specifications

Antigen ODC1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone ODC1/2878R
Conjugate Unconjugated
Description ornithine decarboxylase 1
Gene ODC1
Gene Alias ODC
Host Species Rabbit
Immunogen Recombinant protein corresponding to full-length human ODC1
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 4953
Target Species Human, Mouse, Rat
Content And Storage Store at -20 to -80°C
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.