Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ODF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569988
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569988 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569988 Supplier Invitrogen™ Supplier No. PA569988
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

The outer dense fibers are cytoskeletal structures that surround the axoneme in the middle piece and principal piece of the sperm tail. The fibers function in maintaining the elastic structure and recoil of the sperm tail as well as in protecting the tail from shear forces during epididymal transport and ejaculation. Defects in the outer dense fibers lead to abnormal sperm morphology and infertility. The human outer dense fibers contains at least 10 major proteins and this gene encodes the main protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ODF1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ODF1
Gene Accession No. Q61999
Gene Alias cancer/testis antigen 133; CT133; Heat shock protein beta-10; HSPB10; ODF; Odf1; ODF2; Odf27; ODFP; ODFPG; ODFPGA; ODFPGB; outer dense fiber gene; outer dense fiber of sperm tails 1; outer dense fiber of sperm tails, 27-kD; outer dense fiber protein; outer dense fiber protein 1; outer dense fiber protein-1; Protein RT7; RT7; RTS 5/1; SODF; Sperm outer dense fiber major protein 1
Gene Symbols ODF1
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of mouse ODF1 (aa 117-166).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18285
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.