Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ODF2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | ODF2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
ODF2 Polyclonal specifically detects ODF2 in Human samples. It is validated for Western Blot.Specifications
ODF2 | |
Polyclonal | |
Rabbit | |
Q5BJF6-3 | |
4957 | |
Synthetic peptides corresponding to ODF2(outer dense fiber of sperm tails 2) The peptide sequence was selected from the N terminal of ODF2. Peptide sequence TCTDINTLTRQKELLLQKLSTFEETNRTLRDLLREQHCKEDSERLMEQQG. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
Cenexin, FLJ44866, MGC111096, MGC9034, ODF2/1, ODF2/2, ODF8484-kD, outer dense fiber of sperm tails 2, outer dense fiber protein 2, sperm tail structural protein | |
ODF2 | |
IgG | |
89 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title