Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ODF2 Antibody (SR2297), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | ODF2 |
---|---|
Clone | SR2297 |
Dilution | Western Blot 1:500-1:1000, Flow Cytometry 1:100 |
Applications | Western Blot, Flow Cytometry |
Classification | Monoclonal |
Description
ODF2 Monoclonal antibody specifically detects ODF2 in Human samples. It is validated for Western Blot, Flow CytometrySpecifications
ODF2 | |
Western Blot 1:500-1:1000, Flow Cytometry 1:100 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Cenexin, FLJ44866, MGC111096, MGC9034, ODF2/1, ODF2/2, ODF8484-kD, outer dense fiber of sperm tails 2, outer dense fiber protein 2, sperm tail structural protein | |
A synthesized peptide derived from human ODF2 (Uniprot #: Q5BJF6) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR2297 | |
Western Blot, Flow Cytometry | |
Unconjugated | |
Rabbit | |
Phospho Specific | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
4957 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title