Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ODF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142655
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142655 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142655 Supplier Invitrogen™ Supplier No. PA5142655
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Additional Information: This antibody is expected to recognize reported isoform 1 (NP_002531.3). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.

Seems to be a major component of sperm tail outer dense fibers (ODF). ODFs are filamentous structures located on the outside of the axoneme in the midpiece and principal piece of the mammalian sperm tail and may help to maintain the passive elastic structures and elastic recoil of the sperm tail. May have a modulating influence on sperm motility. Functions as a general scaffold protein that is specifically localized at the distal/subdistal appendages of mother centrioles. Component of the centrosome matrix required for the localization of PLK1 and NIN to the centrosomes. Required for the formation and/or maintenance of normal CETN1 assembly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ODF2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene ODF2
Gene Accession No. A3KGV1
Gene Alias 84 kDa outer dense fiber protein; AI848335; cancer/testis antigen 134; cenexin; cenexin 1; CT134; KKT4; MMTEST29; ODF2; ODF2/1; ODF2/2; Odf84; outer dense fiber of sperm tail 2; outer dense fiber of sperm tails 2; outer dense fiber of sperm tails protein 2; outer dense fiber of sperm tails, 84-kD; outer dense fiber protein 2; RP11-187B3.1; sperm outer dense fiber major protein 2; sperm tail structural protein; testis tissue sperm-binding protein Li 51e; testis-specific autoantigen
Gene Symbols ODF2
Host Species Goat
Immunogen Peptide with sequence C-REKHQASQKENKQ.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18286
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.