Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ODF2 Polyclonal Antibody
ODF2 Polyclonal Antibody
Supplier: Thermo Scientific PA578407
Description
ODF2 Polyclonal Antibody for IHC (P)

Specifications
ODF2 | |
Polyclonal | |
Unconjugated | |
ODF2 | |
84 kDa outer dense fiber protein; AI848335; cancer/testis antigen 134; cenexin; cenexin 1; CT134; KKT4; MMTEST29; ODF2; ODF2/1; ODF2/2; Odf84; outer dense fiber of sperm tail 2; outer dense fiber of sperm tails 2; outer dense fiber of sperm tails protein 2; outer dense fiber of sperm tails, 84-kD; outer dense fiber protein 2; RP11-187B3.1; sperm outer dense fiber major protein 2; sperm tail structural protein; testis tissue sperm-binding protein Li 51e; testis-specific autoantigen | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18286, 4957 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.42 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
A3KGV1, Q5BJF6 | |
ODF2 | |
Recombinant protein encompassing a sequence within the N-terminus region of human ODF2. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction