Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ODF2L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100657
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100657 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100657 Supplier Invitrogen™ Supplier No. PA5100657
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ODF2L.

ODF2L, also named as KIAA1229, has many isoforms with MW 60-80 kDa. The function of ODF2L (Outer dense fiber protein 2-like) remains largely unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ODF2L
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Odf2l
Gene Accession No. Q9D478, Q9ULJ1
Gene Alias 4733401D09Rik; 9630045K08Rik; Basal body centriole-associated protein; Bcap; D3Ertd250e; dJ977L11.1; KIAA1229; ODF2L; outer dense fiber of sperm tails 2 like; outer dense fiber of sperm tails 2-like; outer dense fiber protein 2-like; Protein BCAP; RGD1308397
Gene Symbols Odf2l
Host Species Rabbit
Immunogen A synthesized peptide derived from human ODF2L(Accession Q9ULJ1), corresponding to amino acid residues I100-K150.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 52184, 57489, 685425
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.