Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ODF2L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555751
Description
Immunogen sequence: LLLPLFKDTI EKINFENANL SALNLKISEQ KEILIKELDT FKSVKLALEH LLRKRDYKQT GDNLSSMLLE NLTDNESENT NLKKKVFEKE Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 64%.
ODF2L, also named as KIAA1229, has many isoforms with MW 60-80 kDa. The function of ODF2L (Outer dense fiber protein 2-like) remains largely unknown.
Specifications
ODF2L | |
Polyclonal | |
Unconjugated | |
Odf2l | |
4733401D09Rik; 9630045K08Rik; Basal body centriole-associated protein; Bcap; D3Ertd250e; dJ977L11.1; KIAA1229; ODF2L; outer dense fiber of sperm tails 2 like; outer dense fiber of sperm tails 2-like; outer dense fiber protein 2-like; Protein BCAP; RGD1308397 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57489 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9ULJ1 | |
Odf2l | |
Recombinant protein corresponding to Human ODF2L. Recombinant protein control fragment (Product #RP-94164). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction