Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ODF3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548279
Description
Outer dense fibers are filamentous structures located on the outside of the axoneme in the midpiece and principal piece of the mammalian sperm tail. May help to maintain the passive elastic structures and elastic recoil of the sperm tail.
Specifications
ODF3 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q96PU9, Q920N1 | |
ODF3 | |
KLH conjugated synthetic peptide between 170-199 amino acids from the C-terminal region of human ODF3. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
ODF3 | |
Outer dense fiber protein 3, Outer dense fiber of sperm tails protein 3, Sperm tail protein SHIPPO 1, Transcript induced in spermiogenesis protein 50, ODF3, SHIPPO1, TISP50 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
113746, 69287 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction