Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ODF3B Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989021
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9021 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9021 Supplier Bioss Supplier No. BS9369R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen ODF3B
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ODF3B
Gene Accession No. A8MYP8
Gene Alias ODF3B; ODF3L3; orf2 5′ to PD-ECGF/TP; outer dense fiber of sperm tails 3B; Outer dense fiber protein 3B; Outer dense fiber protein 3-like protein 3
Gene Symbols ODF3B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ODF3B.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 440836
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.