Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ODF3L1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113113
Description
ODF3L1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Odf3l1 is a protein coding gene.
Specifications
| ODF3L1 | |
| Polyclonal | |
| Unconjugated | |
| Odf3l1 | |
| Odf3l1; outer dense fiber of sperm tails 3 like 1; outer dense fiber of sperm tails 3-like 1; outer dense fiber protein 3-like protein 1; RGD1304976 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 161753 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1.5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8IXM7 | |
| Odf3l1 | |
| Recombinant Human Outer dense fiber protein 3-like protein 1 protein (1-274AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction