Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ODF3L2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564353
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564353 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564353 Supplier Invitrogen™ Supplier No. PA564353
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VASPAYSLVR RPSEAPPQDT SPGPIYFLDP KVTRFGRSCT PAYSMQGRAK SRGPEVTPGP Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 85%.

ODF3L2 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ODF3L2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ODF3L2
Gene Accession No. Q3SX64
Gene Alias C19orf19; Gm1157; ODF3L2; outer dense fiber of sperm tails 3 like 2; outer dense fiber of sperm tails 3-like 2; outer dense fiber protein 3-like protein 2; RGD1310260
Gene Symbols ODF3L2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ODF3L2. Recombinant protein control fragment (Product #RP-103663).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284451
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.