Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ODF4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983025
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3025 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3025 Supplier Bioss Supplier No. BS13615R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ODF4 is the component of the outer dense fibers (ODF) of spermatozoa which could be involved in sperm tail structure, sperm movement and general organization of cellular cytoskeleton.This gene encodes a protein that is localized in the outer dense fibers of the tails of mature sperm. This protein is thought to have some important role in the sperm tail. Sequence Note: The RefSeq record was created from transcript and genomic sequence data to make the sequence consistent with the reference genome assembly. The genomic coordinates used for the transcript record were based on transcript alignments.

Specifications

Antigen ODF4
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ODF4
Gene Accession No. Q2M2E3
Gene Alias cancer/testis antigen 134; cancer/testis antigen 136; CT134; CT136; hOPPO1; MGC138215; MGC138219; ODF4; OPPO1; outer dense fiber 4; outer dense fiber of sperm tails 4; Outer dense fiber of sperm tails protein 4; Outer dense fiber protein 4; Testis-specific protein oppo 1
Gene Symbols ODF4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ODF4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146852
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.