Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ODF4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP160018
Description
ODF4 Polyclonal specifically detects ODF4 in Human samples. It is validated for Western Blot.Specifications
ODF4 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
cancer/testis antigen 134, cancer/testis antigen 136, CT134, CT136, hOPPO1, MGC138215, OPPO1MGC138219, outer dense fiber 4, outer dense fiber of sperm tails 4, Outer dense fiber of sperm tails protein 4, outer dense fiber protein 4, Testis-specific protein oppo 1 | |
Rabbit | |
29 kDa | |
100 μL | |
Primary | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q2M2E3 | |
ODF4 | |
Synthetic peptides corresponding to ODF4(outer dense fiber of sperm tails 4) The peptide sequence was selected from the N terminal of ODF4. Peptide sequence MDAEYSGNEFPRSEGERDQHQRPGKERKSGEAGWGTGELGQDGRLLSSTL. | |
Affinity purified | |
RUO | |
146852 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction