Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ODQ, 98%

Catalog No. p-3779366
Click to view available options
Quantity:
10 mg
50 mg
Packaging:
Glass bottle

CAS: 41443-28-1 | C9H5N3O2 | 187.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41443-28-1
Molecular Formula C9H5N3O2
Molecular Weight (g/mol) 187.2
MDL Number MFCD00792620
InChI Key LZMHWZHOZLVYDL-UHFFFAOYSA-N
Synonym 1h-1,2,4 oxadiazolo 4,3-a quinoxalin-1-one, odq, 1,2,4 oxadiazolo 4,3-a quinoxalin-1-one, unii-s57v2nmv38, 1h-1,2,4 oxadiazolo 4,3-a-quinoxalin-1-one, 1h-1,2,4 oxadiazolo-4,3,a quinoxalin-1-one, 1-h-1,2,4 oxadiazolo-4,3,2 quinoxalin-1-ine, 1h-odq
PubChem CID 1456
IUPAC Name [1,2,4]oxadiazolo[4,3-a]quinoxalin-1-one
SMILES C1=CC=C2C(=C1)N=CC3=NOC(=O)N23

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 160°C to 170°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 10 mg
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in chloroform,ethylacetate,dmso
Formula Weight 187.2
Percent Purity 98%
Physical Form Powder
Chemical Name or Material ODQ
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.