Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ODR4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984340
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4340 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4340 Supplier Bioss Supplier No. BS19564R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May play a role in the trafficking of a subset of G-protein coupled receptors.

Specifications

Antigen ODR4
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ODR4
Gene Accession No. Q5SWX8
Gene Alias 1810053E15Rik; BC003331; C1orf27; cDNA sequence BC003331; chromosome 1 open reading frame 27; hODR-4; LAG1-interacting protein; mODR-4; odorant response abnormal 4; Odr4; odr-4; olfactory specific odr-4; Protein odr-4 homolog; RGD1307830; similar to cDNA sequence BC003331; transactivated by recombinant transforming growth factor beta; Transactivated by transforming growth factor beta protein 1; TTG1; TTG1A
Gene Symbols ODR4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ODR4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54953
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.