Description
- For aseptic and cleanroom conditions; designed to be easily cleaned
- Great panoramic view
- High temperature, nonvented and top-vented body options available
- UV-absorbing polycarbonate, durable antifog coating
- Ultra comfortable, soft goggle body for a snug seal
- Easily fits over prescription eyewear
- Made of anallergic materials
- Easy strap adjustment
Order Info
Replacement parts available (See Cat. No. 19-823-051).
Specifications
Specifications
| Lens Tint | Clear |
| Product Type | Cleanroom goggles |
| Frame Color | Blue |
| Ventilation Details | Non-vent |
| Material | Polycarbonate |
| Frame Material | High Temperature TPR |