Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ODZ3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988789
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8789 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8789 Supplier Bioss Supplier No. BS9061R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This protein involved in neural development, regulating the establishment of proper connectivity within the nervous system. It promotes axon guidance and homophilic cell adhesion. And it plays a role in the development of the visual pathway; regulates the formation in ipsilateral retinal mapping to both the dorsal lateral geniculate nucleus (dLGN) and the superior colliculus (SC). May be involved in the differentiation of the fibroblast-like cells in the superficial layer of mandibular condylar cartilage into chondrocytes.

Specifications

Antigen ODZ3
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TENM3
Gene Accession No. Q9P273
Gene Alias 2610100B16Rik; FLJ10474; FLJ10886; KIAA1455; MCOPCB9; mKIAA1455; odd Oz/ten-m homolog 3; odz, odd Oz/ten-m homolog 3; Odz1; ODZ3; ODZ3-like protein; Protein Odd Oz/ten-m homolog 3; Ten-3; Tenascin-M3; Teneurin transmembrane protein 3; Teneurin-3; TENM3; Ten-m3; testicular tissue protein Li 195; TNM3
Gene Symbols TENM3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ODZ3/Teneurin 3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55714
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.