Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OFCC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557375
Description
Immunogen sequence: AEFLMVKEDR EATEGTGNPA FNMSSPDLSA CQTAEKKVIR HDMPDRTLAA HQQKFRLPAS AEPKGNEYGR NY Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 71%.
The function of this protein remains unknown.
Specifications
OFCC1 | |
Polyclonal | |
Unconjugated | |
OFCC1 | |
MRDS1; Ofcc1; ojoplano; opo; orofacial cleft 1 candidate 1; orofacial cleft 1 candidate gene 1 protein homolog; orofacial clefting chromosomal breakpoint region 1; orofacial clefting chromosomal breakpoint region candidate 1 protein homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
266553 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IZS5 | |
OFCC1 | |
Recombinant protein corresponding to Human OFCC1. Recombinant protein control fragment (Product #RP-95014). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction