Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OFCC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113097
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113097 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113097 Supplier Invitrogen™ Supplier No. PA5113097
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OFCC1 Polyclonal Antibody for Western Blot, ELISA

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OFCC1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene OFCC1
Gene Accession No. Q8BGX4, Q8IZS5
Gene Alias MRDS1; Ofcc1; ojoplano; opo; orofacial cleft 1 candidate 1; orofacial cleft 1 candidate gene 1 protein homolog; orofacial clefting chromosomal breakpoint region 1; orofacial clefting chromosomal breakpoint region candidate 1 protein homolog
Gene Symbols OFCC1
Host Species Rabbit
Immunogen Recombinant Human Orofacial cleft 1 candidate gene 1 protein (3-231AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 218165, 266553
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.