Learn More
Invitrogen™ OFD1 Polyclonal Antibody

Description
Recommended positive controls: Molt-4. Predicted reactivity: Bovine (80%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | OFD1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1.21 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene | OFD1 |
| Gene Accession No. | O75665 |
| Gene Alias | 71-7A; CXorf5; DXGgc7e; JBTS10; Ofd1; OFD1 centriole and centriolar satellite protein; ofd1 protein; OFD1, centriole and centriolar satellite protein; oral-facial-digital syndrome 1; oral-facial-digital syndrome 1 gene homolog; oral-facial-digital syndrome 1 gene homolog (human); oral-facial-digital syndrome 1 protein; Oral-facial-digital syndrome 1 protein homolog; ORF2; Protein 71-7A; putative oral-facial-digital syndrome 1 protein; RGD1562231; RP23; SGBS2; wu:fa07a10; wu:fd17e11; zgc:92562 |
| Gene Symbols | OFD1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.