Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OFD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570403
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570403 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570403 Supplier Invitrogen™ Supplier No. PA570403
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 79%.

This gene is located on the X chromosome and encodes a centrosomal protein. A knockout mouse model has been used to study the effect of mutations in this gene. The mouse gene is also located on the X chromosome, however, unlike the human gene it is not subject to X inactivation. Mutations in this gene are associated with oral-facial-digital syndrome type I and Simpson-Golabi-Behmel syndrome type 2. Many pseudogenes have been identified; a single pseudogene is found on chromosome 5 while as many as fifteen have been found on the Y chromosome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OFD1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene OFD1
Gene Accession No. 0
Gene Alias 71-7A; CXorf5; DXGgc7e; JBTS10; Ofd1; OFD1 centriole and centriolar satellite protein; ofd1 protein; OFD1, centriole and centriolar satellite protein; oral-facial-digital syndrome 1; oral-facial-digital syndrome 1 gene homolog; oral-facial-digital syndrome 1 gene homolog (human); oral-facial-digital syndrome 1 protein; Oral-facial-digital syndrome 1 protein homolog; ORF2; Protein 71-7A; putative oral-facial-digital syndrome 1 protein; RGD1562231; RP23; SGBS2; wu:fa07a10; wu:fd17e11; zgc:92562
Gene Symbols OFD1
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human OFD1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 302661
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.