Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OGDH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582681
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582681 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582681 Supplier Invitrogen™ Supplier No. PA582681
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LRTCAAKLRP LTASQTVKTF SQNRPAAART FQQIRCYSAP VAAEPFLSGT SSNYVEEMYC AWLENPKSV Highest antigen sequence indentity to the following orthologs: Mouse - 97%, Rat - 97%.

This gene encodes one subunit of the 2-oxoglutarate dehydrogenase complex. This complex catalyzes the overall conversion of 2-oxoglutarate (alpha-ketoglutarate) to succinyl-CoA and CO(2) during the Krebs cycle. The protein is located in the mitocondrial matrix and uses thiamine pyrophosphate as a cofactor. A congential deficiency in 2-oxoglutarate dehydrogenase activity is believed to lead to hypotonia, metabolic acidosis, and hyperlactatemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OGDH
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OGDH
Gene Accession No. Q02218
Gene Alias 2210403E04Rik; 2210412K19Rik; 2-oxoglutarate dehydrogenase complex component E1; 2-oxoglutarate dehydrogenase E1 component, mitochondrial; 2-oxoglutarate dehydrogenase, mitochondrial; AA409584; AKGDH; Alpha-ketoglutarate dehydrogenase; d1401; E1k; Kiaa4192; mKIAA4192; OGDC; OGDC-E1; Ogdh; Ogdhl; oxoglutarate (alpha-ketoglutarate) dehydrogenase (lipoamide); oxoglutarate decarboxylase; oxoglutarate dehydrogenase; oxoglutarate dehydrogenase (lipoamide); oxoglutarate dehydrogenase (succinyl-transferring); testicular tissue protein Li 131
Gene Symbols OGDH
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human OGDH. Recombinant protein control fragment (Product #RP-92648).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4967
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.