Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OGDHL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100659
Description
Antibody detects endogenous levels of total OGDHL.
The function of OGDHL remains unknown.
Specifications
OGDHL | |
Polyclonal | |
Unconjugated | |
OGDHL | |
2-oxoglutarate dehydrogenase complex component E1-like; 2-oxoglutarate dehydrogenase-like, mitochondrial; 2-oxoglutarate dehydrogenase-like, mitochondrial-like; alpha-ketoglutarate dehydrogenase-like; KIAA1290; LOW QUALITY PROTEIN: 2-oxoglutarate dehydrogenase-like, mitochondrial; OGDC-E1-like; OGDHL; oxoglutarate dehydrogenase L; oxoglutarate dehydrogenase like; oxoglutarate dehydrogenase-like | |
Rabbit | |
Affinity Chromatography | |
RUO | |
239017, 290566, 55753 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9ULD0 | |
OGDHL | |
A synthesized peptide derived from human OGDHL(Accession Q9ULD0), corresponding to amino acid residues R559-S609. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction