Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OGDHL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA544716
Description
Peptide sequence: VFGWRSRSSG PPATFPSSKG GGGSSYMEEM YFAWLENPQS VHKSWDSFFR Sequence homology: Cow: 85%; Dog: 100%; Horse: 92%; Human: 100%; Pig: 100%; Rat: 85%.
The function of OGDHL remains unknown.
Specifications
OGDHL | |
Polyclonal | |
Unconjugated | |
OGDHL | |
2-oxoglutarate dehydrogenase complex component E1-like; 2-oxoglutarate dehydrogenase-like, mitochondrial; 2-oxoglutarate dehydrogenase-like, mitochondrial-like; alpha-ketoglutarate dehydrogenase-like; KIAA1290; LOW QUALITY PROTEIN: 2-oxoglutarate dehydrogenase-like, mitochondrial; OGDC-E1-like; OGDHL; oxoglutarate dehydrogenase L; oxoglutarate dehydrogenase like; oxoglutarate dehydrogenase-like | |
Rabbit | |
Affinity Chromatography | |
RUO | |
55753 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9ULD0 | |
OGDHL | |
Synthetic peptide directed towards the N-terminal of human OGDHL (aa 26-75). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction