Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Ogerin
GREENER_CHOICESDP

Catalog No. 572250 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-225-0 50 mg
57-221-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 57-225-0 Supplier Tocris Bioscience™ Supplier No. 5722/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective positive allosteric modulator at GPR68

Ogerin is a selective positive allosteric modulator (PAM) of GPR68 (pEC50 = 6.83), which exhibits selectivity for GPR68 over 58 other GPCRs. Ogerin attenuates contextual-based fear memory in mice.

Specifications

Chemical Name or Material 2-[4-Amino-6-[(phenylmethyl)amino]-1,3,5-triazin-2-yl]benzenemethanol
CAS 1309198-71-7
Quantity 50 mg
Target Orphan 7-TM Receptor Agonists
Molecular Formula C17H17N5O
Formula Weight 307.35
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.