Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OGFOD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110058
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110058 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110058 Supplier Invitrogen™ Supplier No. PA5110058
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MNGKRPAEPG PARVGKKGKK EVMAEFSDAV TEETLKKQVA EAWSRRTPFS HEVIVMDMDP FLHCVIPNFI QSQDFLEGLQ KELMNLDFHE KYNDLYKFQQ SDDLKKRREP HISTLRKILF EDFRSWLSDI SKIDLESTID.

OGFOD1, a member of the 2-OG-Fe(II) dioxygenase family, plays an important role in ischemic survival and stress. It is an oxidoreductase containing a Prolyl 4-hydroxylase alpha subunit. An OGFOD1 iron binding residue is required for ATPAF1 gene expression. As a novel stress granule component, OGFOD1 plays important proapoptotic roles in the regulation of translation and HRI-mediated phosphorylation of eIF2alpha in cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OGFOD1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene OGFOD1
Gene Accession No. Q8N543
Gene Alias 2-oxoglutarate and iron dependent oxygenase domain containing 1; 2-oxoglutarate and iron-dependent oxygenase domain containing 1; 2-oxoglutarate and iron-dependent oxygenase domain-containing protein 1; 4930415J21Rik; AA387199; AA939912; AW061076; Kiaa1612; mKIAA1612; OGFOD1; Prolyl 3-hydroxylase OGFOD1; RGD1308848; Termination and polyadenylation 1 homolog; TPA1; TPA1, termination and polyadenylation 1, homolog; uS12 prolyl 3-hydroxylase
Gene Symbols OGFOD1
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55239
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.