Learn More
Invitrogen™ OGFOD1 Polyclonal Antibody

Description
A suggested positive control is Hela cell lysate. PA5-21100 can be used with blocking peptide PEP-1214.
Specifications
Specifications
| Antigen | OGFOD1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | OGFOD1 |
| Gene Accession No. | Q3U0K8, Q8N543 |
| Gene Alias | 2-oxoglutarate and iron dependent oxygenase domain containing 1; 2-oxoglutarate and iron-dependent oxygenase domain containing 1; 2-oxoglutarate and iron-dependent oxygenase domain-containing protein 1; 4930415J21Rik; AA387199; AA939912; AW061076; Kiaa1612; mKIAA1612; OGFOD1; Prolyl 3-hydroxylase OGFOD1; RGD1308848; Termination and polyadenylation 1 homolog; TPA1; TPA1, termination and polyadenylation 1, homolog; uS12 prolyl 3-hydroxylase |
| Gene Symbols | OGFOD1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.