Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OGFOD2 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176533MFV500
Description
OGFOD2 Polyclonal antibody specifically detects OGFOD2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISASpecifications
OGFOD2 | |
Polyclonal | |
Western Blot, ELISA | |
2-oxoglutarate and iron-dependent oxygenase domain containing 2, DKFZp686H15154, EC 1.14.11.-, FLJ13491, FLJ37501, MGC120434, MGC120436,2-oxoglutarate and iron-dependent oxygenase domain-containing protein 2 | |
Antibody was raised against a 17 amino acid synthetic peptide near the center of human OGFOD2. The immunogen is located within amino acids 140 - 190 of OGFOD2. Amino Acid Squence: EHFEQSDMPKGRPNTMN | |
0.1 mL | |
Apoptosis | |
79676 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction