Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OGFOD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558790
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558790 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558790 Supplier Invitrogen™ Supplier No. PA558790
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TALTEPLEVE HVVGQGVLHR GGQLHGARPL GTGERWNLVV WLRASAVRNS LCPMCCREPD LVDDEGFGDG FTREEPATVD VCA Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 88%.

OGFOD2 is a member of the 2-OG-Fe(II) dioxygenase family which plays an important role in ischemic survival and stress. Like the related protein OGFOD1, OGFOD2 is an oxidoreductase containing a Prolyl 4-hydroxylase alpha subunit. Little is known about the function of OGFOD2, but its similarity to OGFOD1 suggests that it is a novel stress granule component that may play an important proapoptotic role in the regulation of translation and HRI-mediated phosphorylation of eIF2alpha in cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OGFOD2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OGFOD2
Gene Accession No. Q6N063
Gene Alias 1300006G11Rik; 2-oxoglutarate and iron dependent oxygenase domain containing 2; 2-oxoglutarate and iron-dependent oxygenase domain containing 2; 2-oxoglutarate and iron-dependent oxygenase domain-containing protein 2; 5730405M13Rik; AU041636; AW552395; OGFOD2; RGD1310594
Gene Symbols OGFOD2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human OGFOD2. Recombinant protein control fragment (Product #RP-97692).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79676
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.